Learn More
AdipoGen Zmp1 M tuberculosis His
SDP
Catalog No. 502562764
Encompass_Preferred
Protein. Full-length Zmp1 Mycobacterium tuberculosis is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content N/A. Liquid. In 10mM TRIS-HCl, pH 8.0. Prefered cleavage sequence is PxxF. Purity >98% SDS-PAGE. M13 endopeptidases are single-pass, transmembrane zinc-metallopeptidases responsible in human for the regulation of the biological activity of many hormones and peptides and are involved in many important processes such as blood pressure regulation Neprilysin or NEP1, cardiovascular development Endothelin Converting Enzyme-1 or ECE-12, prevention of haemolytic reaction KELL and phosphate homeostasis PHEX. Zmp1 is a soluble enzyme member of the M13 endopeptidases family that plays a key role in Mycobacterium tuberculosis pathogenicity with a still unknown mechanism.
Catalog No. 50-256-2764 Supplier AdipoGen Supplier No. AG40T0103C050
Protein. Full-length Zmp1 Mycobacterium tuberculosis is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content N/A. Liquid. In 10mM TRIS-HCl, pH 8.0. Prefered cleavage sequence is PxxF. Purity >98% SDS-PAGE. M13 endopeptidases are single-pass, transmembrane zinc-metallopeptidases responsible in human for the regulation of the biological activity of many hormones and peptides and are involved in many important processes such as blood pressure regulation Neprilysin or NEP1, cardiovascular development Endothelin Converting Enzyme-1 or ECE-12, prevention of haemolytic reaction KELL and phosphate homeostasis PHEX. Zmp1 is a soluble enzyme member of the M13 endopeptidases family that plays a key role in Mycobacterium tuberculosis pathogenicity with a still unknown mechanism.