Learn More
Aviva Systems Biology Corp ZMPSTE24 Antibody - N-terminal region (ARP46665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMPSTE24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZMPSTE24. Alternate names: FACE-1, FACE1, FLJ14968, STE24, Ste24p, HGPS, PRO1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59373