Learn More
Aviva Systems Biology Corp ZMYM3 Antibody - N-terminal region (ARP34515_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34515_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMYM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZMYM3. Alternate names: DXS6673E, KIAA0385, MYM, XFIM, ZNF198L2, ZNF261ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52962