Learn More
Aviva Systems Biology Corp ZMYM4 Antibody - N-terminal region (ARP34210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMYM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZMYM4. Alternate names: CDIR, DKFZp686B09210, DKFZp686L0547, KIAA0425, MYM, ZNF198L3, ZNF262ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55300