Learn More
Aviva Systems Biology Corp ZMYND11 Antibody - N-terminal region (ARP34705_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34705_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMYND11. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZMYND11. Alternate names: BRAM1, BS69, MGC111056, RP11-486H9.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55523