Learn More
Aviva Systems Biology Corp ZMYND12 Antibody - middle region (ARP50709_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50709_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMYND12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZMYND12. Alternate names: DKFZp434N2435ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60416