Learn More
Aviva Systems Biology Corp ZMYND17 Antibody - C-terminal region (ARP50392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMYND17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZMYND17. Alternate names: FLJ39565, ZMYND17ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-53629