Learn More
Aviva Systems Biology Corp ZMYND19 Antibody - middle region (ARP50773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMYND19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZMYND19. Alternate names: MIZIP, RP11-48C7.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60430