Learn More
Bioworld ZN-CHELATED SEPAROPOREAGA
Supplier:  Bioworld 201810492
Encompass
ZN-CHELATED SEPAROPORE (AGAROSE): Zn-Chelated Separopore is an imminodiacetate (IDA) immobilized metal affinity chromatography (IMAC) matrix that seperates proteins via their differences in affinity toZn metal ions. Properties: Affinity...
Catalog No. 50-197-791