Learn More
Medchemexpress LLC Zn(II)-EDTA disodium tetrahydrate | 176736-49-5 | MFCD00151728 | 95.0% | 471.63 g/mol | C10H20N2Na2O12Zn | 500 G
SDP
Catalog No. 5000283639
Encompass_Preferred
Zn(II)-EDTA disodium tetrahydrate is a zinc chelate reagent supplied as a crystalline solid tetrahydrate for research and analytical use. It functions as a metal-chelating agent in biochemical assays, sample preparation, and analytical workflows where coordinated zinc complexes are required.
  • Metal chelating agent for zinc complexation.
  • Suitable for biochemical assays and analytical applications.
  • Supplied as a stable solid tetrahydrate for easy weighing and storage.
  • Typical purity 95.0%, appropriate for research use.
  • Available in laboratory pack sizes including 500 G.
Catalog No. 50-002-83639 Supplier Medchemexpress LLC Supplier No. HYW127776500G
Zn(II)-EDTA disodium tetrahydrate is a zinc chelate reagent supplied as a crystalline solid tetrahydrate for research and analytical use. It functions as a metal-chelating agent in biochemical assays, sample preparation, and analytical workflows where coordinated zinc complexes are required.
  • Metal chelating agent for zinc complexation.
  • Suitable for biochemical assays and analytical applications.
  • Supplied as a stable solid tetrahydrate for easy weighing and storage.
  • Typical purity 95.0%, appropriate for research use.
  • Available in laboratory pack sizes including 500 G.