Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 250mg, Zn(II) meso-Tetraphenylporphine (1-3% chlorin), >95%, [14074-80-7], T40942 678.129 C44H28N4Zn

Catalog No. 501456185
Encompass_Preferred

Compound: Zn(II) meso-Tetraphenylporphine (1-3% chlorin)

CAS#: 14074-80-7

MDL#: not listed

Background: A synthetic zinc(II) tetraphenylporphyrin complex used as a reference standard and building block in porphyrin chemistry, materials research, and photophysical studies of metalloporphyrins.

Storage: Store at room temperature, protected from light.

Catalog No. 50-145-6185 Supplier Frontier Specialty Chemicals Supplier No. T40942250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Zn(II) meso-Tetraphenylporphine (1-3% chlorin)

CAS#: 14074-80-7

MDL#: not listed

Background: A synthetic zinc(II) tetraphenylporphyrin complex used as a reference standard and building block in porphyrin chemistry, materials research, and photophysical studies of metalloporphyrins.

Storage: Store at room temperature, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.