Learn More
Bioworld Zn-IMAC 5 x 2mL Columns, 5 Column
Catalog No. 50197635
Encompass_Preferred
Zn-immobilized metal affinity chromatography (IMAC) columns consist of Zn-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Zn-IMAC Columns separate proteins via their differences in affinity toZn2+ metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged withZn metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 4BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Zn2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.
Catalog No. 50-197-635 Supplier Bioworld Supplier No. 201700151
Zn-immobilized metal affinity chromatography (IMAC) columns consist of Zn-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Zn-IMAC Columns separate proteins via their differences in affinity toZn2+ metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged withZn metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 4BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Zn2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.