Learn More
Echelon Biosciences Research Labs ZN PROTOPORPHYRIN IX 100 mg
SDP
Catalog No. 50720333
Encompass
Zn (II) Protoporphyrin IX is an inhibitor of heme oxygenase (the enzyme that catalyzes the conversion of heme to biliverdin in the heme degradation pathway) and guanylyl cyclase. Heme oxygenase has been implicated in tumor cell resistance to chemotherapy, reduction of free radical formation and inflammation, and associated with vascular repair.h4>StorageZinc (II) Protoporphyrin IX is stable for at least one year when stored as a solid, protected from moisture, at -20 C. Protect from light. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-333 Supplier Echelon Biosciences Research Labs Supplier No. FH060100MG
Zn (II) Protoporphyrin IX is an inhibitor of heme oxygenase (the enzyme that catalyzes the conversion of heme to biliverdin in the heme degradation pathway) and guanylyl cyclase. Heme oxygenase has been implicated in tumor cell resistance to chemotherapy, reduction of free radical formation and inflammation, and associated with vascular repair.h4>StorageZinc (II) Protoporphyrin IX is stable for at least one year when stored as a solid, protected from moisture, at -20 C. Protect from light. This item is non returnable