Learn More
AdipoGen ZnAF-1 Solution
SDP
Supplier:  AdipoGen CDXZ0505M001
Encompass_Preferred
Chemical. CAS 321859-09-0. Formula C34H28N4O5. MW 572.61. Synthetic. A membrane-impermeable ethylenediaminofluoresceinyl compound that acts as a high-affinity Zn2-specific fluorescent probe. Spectral data lambdaex 492 nm lambdaem 514 nm in PBS.
Catalog No. 50-255-9861