Learn More
AdipoGen ZnAF-1F DA
SDP
Catalog No. 502559875
Encompass_Preferred
Chemical. Formula C38H30F2N4O7. MW 692.66. Synthetic. For application to living cells, the diacetyl derivative of ZnAF-1F, ZnAF-1F DA, was synthesized. ZnAF-1F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-1F, which is retained in the cells. ZnAF-1F DA has been used to measure the changes of intracellular Zn2 in cultured cells and hippocampal slices. Spectral data lambdaex 492nm lambdaem 514nm in PBS.
Catalog No. 50-255-9875 Supplier AdipoGen Supplier No. CDXZ0011M005
Chemical. Formula C38H30F2N4O7. MW 692.66. Synthetic. For application to living cells, the diacetyl derivative of ZnAF-1F, ZnAF-1F DA, was synthesized. ZnAF-1F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-1F, which is retained in the cells. ZnAF-1F DA has been used to measure the changes of intracellular Zn2 in cultured cells and hippocampal slices. Spectral data lambdaex 492nm lambdaem 514nm in PBS.