Learn More
AdipoGen ZnAF-2
SDP
Supplier:  AdipoGen CDXZ0006M005
Encompass_Preferred
Chemical. CAS 321859-11-4. Formula C34H28N4O5. MW 572.61. Synthetic. A membrane-impermeable ethylenediaminofluoresceinyl compound that acts as a high-affinity Zn2-specific fluorescent probe Kd = 2.7 nM. It shows low basal fluorescence and the fluorescence intensity increases ~51-fold upon stoichiometric 11 binding to Zn2. Exhibits little affinity towards Ca2, Mg2, Na, or K. Spectral data lambdaex 492nm lambdaem 515nm in PBS.
Catalog No. 50-255-9889