Learn More
Enzo Life Sciences ZnAF-2 DA (1mg). CAS: 357339-96-9
SDP
Catalog No. 502009607
Encompass_Preferred
Cell permeable derivative of ZnAF-2 (ALX-620-072). Fluorescent reagent (Ex(max): 492nm; Em(max): 514nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2.7nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion. Alternative name: 6-[N-[N',N'-bis(2-Pyridinylmethyl)-2-aminoethyl]amino-3',6'-bis(acetyloxy)-spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one. Formulation: Dissolved in 0.3ml DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9607 Supplier Enzo Life Sciences Supplier No. ALX620076M001
Cell permeable derivative of ZnAF-2 (ALX-620-072). Fluorescent reagent (Ex(max): 492nm; Em(max): 514nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2.7nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion. Alternative name: 6-[N-[N',N'-bis(2-Pyridinylmethyl)-2-aminoethyl]amino-3',6'-bis(acetyloxy)-spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one. Formulation: Dissolved in 0.3ml DMSO. Long Term Storage: -20°C.