Learn More
AdipoGen ZnAF-2 DA
SDP
Supplier:  AdipoGen CDXZ0008M005
Encompass_Preferred
Chemical. CAS 357339-96-9. Formula C38H32N4O7. MW 656.68. Synthetic. Suitable for the fluorimetric detection of Zn2 after esterase hydrolysis. Spectral data lambdaex 492nm lambdaem 515nm in PBS.
Catalog No. 50-255-9883