Learn More
AdipoGen ZnAF-2F DA
SDP
Catalog No. 502559873
Encompass_Preferred
Chemical. CAS 443302-10-1. Formula C38H30F2N4O7. MW 692.66. Synthetic. For application to living cells, the diacetyl derivative of ZnAF-2F, ZnAF-2F DA, was synthesized. ZnAF-2F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-2F, which is retained in the cells. Using ZnAF-2F DA we could measure the changes of intracellular Zn2 in cultured cells and hippocampal slices. Spectral data lambdaex 492nm lambdaem 515nm in PBS.
Catalog No. 50-255-9873 Supplier AdipoGen Supplier No. CDXZ0012M005
Chemical. CAS 443302-10-1. Formula C38H30F2N4O7. MW 692.66. Synthetic. For application to living cells, the diacetyl derivative of ZnAF-2F, ZnAF-2F DA, was synthesized. ZnAF-2F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-2F, which is retained in the cells. Using ZnAF-2F DA we could measure the changes of intracellular Zn2 in cultured cells and hippocampal slices. Spectral data lambdaex 492nm lambdaem 515nm in PBS.