Learn More
Aviva Systems Biology Corp ZNF10 Antibody - N-terminal region (P100847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF10. Alternate names: KOX1ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-62741