Learn More
Aviva Systems Biology Corp ZNF101 Antibody - N-terminal region (ARP36099_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36099_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF101. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF101. Alternate names: DKFZp570I0164, HZF12, MGC149565, MGC149566ApplicationWB. Species reactivity: Human
Catalog No. 50-284-482