Learn More
Aviva Systems Biology Corp ZNF117 Antibody - N-terminal region (ARP47632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF117. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF117. Alternate names: H-plk, HPF9, MGC22613ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59616