Learn More
Aviva Systems Biology Corp ZNF12 Antibody - N-terminal region (ARP37766_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37766_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF12. Alternate names: KOX3, HZF11, GIOT-3, ZNF325ApplicationWB. Species reactivity: Dog, Horse, Human, Rat
Catalog No. 50-014-56426