Learn More
Aviva Systems Biology Corp ZNF121 Antibody - middle region (ARP50353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF121. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF121. Alternate names: D19S204, ZHC32, ZNF20ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60323