Learn More
Aviva Systems Biology Corp ZNF124 Antibody - N-terminal region (ARP38327_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38327_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF124. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF124. Alternate names: ZK7, HZF16, HZF-16ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56608