Learn More
Aviva Systems Biology Corp ZNF124 Antibody - middle region (ARP35670_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35670_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF124. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF124. Alternate names: HZF-16, HZF16, MGC117046, ZK7ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55811