Learn More
Aviva Systems Biology Corp ZNF133 Antibody - N-terminal region (ARP35674_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35674_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF133. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF133. Alternate names: ZNF150, pHZ-13, pHZ-66ApplicationIF, WB. Species reactivity: Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-55813