Learn More
Aviva Systems Biology Corp ZNF134 Antibody - N-terminal region (ARP35676_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35676_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF134. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF134. Alternate names: pHZ-15ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55815