Learn More
Aviva Systems Biology Corp ZNF14 Antibody - N-terminal region (ARP51595_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51595_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF14. Alternate names: GIOT-4, KOX6ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60568