Learn More
Aviva Systems Biology Corp ZNF140 Antibody - middle region (ARP38336_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38336_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF140. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF140. Alternate names: pHZ-39ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rat
Catalog No. 50-014-56612