Learn More
Aviva Systems Biology Corp ZNF141 Antibody - N-terminal region (ARP37726_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37726_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF141. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF141. Alternate names: D4S90, pHZ-44ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56408