Learn More
Aviva Systems Biology Corp ZNF142 Antibody - C-terminal region (ARP38554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF142. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF142. Alternate names: pHZ-49ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-743