Learn More
Aviva Systems Biology Corp ZNF146 Antibody - middle region (ARP35828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF146. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF146. Alternate names: MGC125660, MGC125661, OZFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55881