Learn More
Aviva Systems Biology Corp ZNF148 Antibody - middle region (ARP34536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF148. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF148. Alternate names: BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55438