Learn More
Aviva Systems Biology Corp ZNF154 Antibody - N-terminal region (ARP47779_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47779_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF154. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF154. Alternate names: MGC176628, MGC176661, pHZ-92ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-014-59666