Learn More
Aviva Systems Biology Corp ZNF16 Antibody - N-terminal region (ARP38884_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38884_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF16. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF16. Alternate names: KOX9ApplicationIHC, WB. Species reactivity: Human, Mouse, Pig
Catalog No. 50-014-56833