Learn More
Aviva Systems Biology Corp ZNF165 Antibody - middle region (ARP35689_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35689_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF165. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF165. Alternate names: LD65, ZSCAN7, CT53ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-55819