Learn More
Aviva Systems Biology Corp ZNF167 Antibody - N-terminal region (ARP35958_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35958_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF167. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF167. Alternate names: ZFP, ZNF64, ZNF448, ZKSCAN7, ZNF167ApplicationIHC, WB. Species reactivity: Human, Rat
Catalog No. 50-014-55945