Learn More
Aviva Systems Biology Corp ZNF169 Antibody - N-terminal region (ARP47749_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47749_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF169. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF169. Alternate names: MGC51961ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-59657