Learn More
Aviva Systems Biology Corp ZNF174 Antibody - N-terminal region (P100902_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100902_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF174. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF174. Alternate names: ZSCAN8ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit
Catalog No. 50-014-62777