Learn More
Aviva Systems Biology Corp ZNF175 Antibody - N-terminal region (ARP35832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF175. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF175. Alternate names: OTK18ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55882