Learn More
Aviva Systems Biology Corp ZNF177 Antibody - N-terminal region (ARP38346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF177. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF177. Alternate names: PIGXApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-56615