Learn More
Aviva Systems Biology Corp ZNF179 Antibody - middle region (ARP34127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF179. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF179. Alternate names: BFP, RNF112, ZNF179ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55259