Learn More
Aviva Systems Biology Corp ZNF180 Antibody - middle region (ARP35862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF180. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF180. Alternate names: HHZ168ApplicationWB. Species reactivity: Dog, Horse, Human, Rat
Catalog No. 50-014-55894