Learn More
Aviva Systems Biology Corp ZNF182 Antibody - middle region (ARP50546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF182. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF182. Alternate names: HHZ150, KOX14, MGC125383, MGC131713, ZNF21, Zfp182ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62886