Learn More
Aviva Systems Biology Corp ZNF185 Antibody - N-terminal region (ARP38914_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38914_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF185. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF185. Alternate names: -ApplicationWB. Species reactivity: Horse, Human, Mouse
Catalog No. 50-014-56846