Learn More
Aviva Systems Biology Corp ZNF187 Antibody - N-terminal region (ARP31230_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31230_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF187. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human ZNF187. Alternate names: MGC2815, SRE-ZBP, ZSCAN26, ZNF187ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-54187