Learn More
Aviva Systems Biology Corp ZNF189 Antibody - N-terminal region (ARP38348_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38348_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF189. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF189. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-284-730