Learn More
Aviva Systems Biology Corp ZNF189 Antibody - middle region (ARP33547_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF189. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF189. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55063