Learn More
Aviva Systems Biology Corp ZNF19 Antibody - N-terminal region (ARP42331_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42331_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF19. Alternate names: KOX12, MGC51021ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-58186